siRNA Details
| Gene Symbol: Sult1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023135.2 | NP_075624.2 | 4 | 436 | ||
| GenBank mRNA | AK015750.1 | - | 4 | 420 | ||
| S78182.1 | - | 4 | 419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1e1| Gene Symbol: Sult1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023135.2 | NP_075624.2 | 4 | 436 | ||
| GenBank mRNA | AK015750.1 | - | 4 | 420 | ||
| S78182.1 | - | 4 | 419 | |||