siRNA Details
| Gene Symbol: Stfa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025288.2 | NP_079564.1 | 2 | 164 | ||
| GenBank mRNA | AK014349.1 | - | 2 | 167 | ||
| AY169236.1 | - | 2 | 147 | |||
| BC107224.1 | - | 2 | 158 | 40051891 | ||
| BC107225.1 | - | 2 | 160 | 40051892 | ||
| M92419.1 | - | 2 | 148 | |||

Stfa3
Genomic Map