siRNA Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009288.2 | NP_033314.2 | 9 | 1405 | ||
| GenBank mRNA | AK148118.1 | - | 9 | 1348 | ||
| AK220448.1 | - | 9 | 1327 | |||
| BC069840.1 | - | 9 | 1478 | |||
| BC096624.1 | - | 9 | 1490 | |||
| BC128363.1 | - | 9 | 1291 | 40111177 | ||
| D89728.1 | - | 9 | 1366 | |||

Stk10
Genomic Map