siRNA Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009288.2 | NP_033314.2 | 1 | 184 | ||
| GenBank mRNA | AK148118.1 | - | 1 | 127 | ||
| AK220448.1 | - | 1 | 106 | |||
| BC069840.1 | - | 1 | 129 | |||
| BC096624.1 | - | 1 | 141 | |||
| BC128363.1 | - | 1 | 70 | 40111177 | ||
| BY207613.1 | - | 1 | 185 | |||
| D89728.1 | - | 1 | 145 | |||

Stk10
Genomic Map