siRNA Details
| Gene Symbol: Plk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011495.2 | NP_035625.2 | 5 | 1141 | ||
| NM_173169.2 | NP_775261.2 | 5 | 1141 | |||
| GenBank mRNA | AK006827.1 | - | 5 | 1047 | ||
| AK045082.1 | - | 5 | 1065 | |||
| AK137080.1 | - | 5 | 1053 | |||
| AK137471.1 | - | 5 | 1052 | |||
| AK164465.1 | - | 5 | 1047 | |||
| BC026785.1 | - | 5 | 1051 | 5150173 | ||
| BC057940.1 | - | 5 | 1038 | 5323295 | ||
| L29479.1 | - | 5 | 1043 | |||
| L29480.1 | - | 5 | 1043 | |||

Plk4
Genomic Map