siRNA Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164639.1 | NP_001158111.1 | 12 | 2787 | ||
| NM_009289.3 | NP_033315.2 | 12 | 2787 | |||
| GenBank mRNA | AF039574.1 | - | 12 | 2792 | ||
| AF112855.1 | - | 12 | 2676 | |||
| AK037798.1 | - | 4 | 761 | |||
| AK122218.1 | - | 12 | 3128 | |||
| BC131674.1 | - | 12 | 2688 | 40110915 | ||
| BC131675.1 | - | 12 | 2688 | 40110921 | ||
| CA535951.1 | - | 2 | 152 | |||
| CF748583.1 | - | 4 | 635 | |||

Slk
Genomic Map