siRNA Details
| Gene Symbol: Aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011496.1 | NP_035626.1 | 5 | 787 | ||
| GenBank mRNA | AK075951.1 | - | 5 | 753 | ||
| AK132006.1 | - | 5 | 781 | |||
| AK182803.1 | - | 2 | 185 | |||
| AK184704.1 | - | 1 | 325 | |||
| BC003261.1 | - | 5 | 779 | 3501444 | ||
| D21099.1 | - | 5 | 652 | |||
| U69107.1 | - | 730 | ||||

Aurkb
Genomic Map