siRNA Details
| Gene Symbol: Sult1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018751.2 | NP_061221.2 | 7 | 782 | ||
| GenBank mRNA | AF033653.1 | - | 7 | 793 | ||
| BC045149.1 | - | 7 | 782 | 6308175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c1| Gene Symbol: Sult1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018751.2 | NP_061221.2 | 7 | 782 | ||
| GenBank mRNA | AF033653.1 | - | 7 | 793 | ||
| BC045149.1 | - | 7 | 782 | 6308175 | ||