siRNA Details
| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016665.2 | NP_057874.2 | 5 | 613 | ||
| GenBank mRNA | AK076181.1 | - | 4 | 672 | ||
| AK080500.1 | - | 5 | 585 | |||
| AK170126.1 | - | 5 | 585 | |||
| BC052335.1 | - | 6 | 863 | |||
| CA320983.1 | - | 4 | 552 | |||
| CJ236047.1 | - | 4 | 525 | |||
| U95003.1 | - | 6 | 676 | |||
| U95004.1 | - | 5 | 577 | |||
| U95005.1 | - | 6 | 850 | |||

Stra13
Genomic Map