siRNA Details
| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016665.2 | NP_057874.2 | 5 | 475 | ||
| GenBank mRNA | AK076181.1 | - | 4 | 534 | ||
| AK080500.1 | - | 5 | 447 | |||
| AK170126.1 | - | 5 | 447 | |||
| BC052335.1 | - | 6 | 725 | |||
| CA320983.1 | - | 4 | 414 | |||
| CJ236047.1 | - | 4 | 387 | |||
| U95003.1 | - | 6 | 539 | |||
| U95005.1 | - | 6 | 713 | |||

Stra13
Genomic Map