siRNA Details
| Gene Symbol: Stra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162475.1 | NP_001155947.1 | 13 | 1228 | ||
| NM_001162476.1 | NP_001155948.1 | 13 | 1539 | |||
| NM_001162479.1 | NP_001155951.1 | 13 | 1333 | |||
| NM_009291.2 | NP_033317.2 | 13 | 1498 | |||
| GenBank mRNA | AF062476.1 | - | 13 | 1498 | ||
| BC075657.1 | - | 13 | 1228 | 30605532 | ||
| CF735552.1 | - | 5 | 398 | |||

Stra6
Genomic Map