siRNA Details
| Gene Symbol: INPP5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321042.1 | NP_001307971.1 | 4, 5 | 908 | ||
| NM_005539.4 | NP_005530.3 | 7, 8 | 855 | |||
| XM_017016204.1 | XP_016871693.1 | 6, 7 | 509 | |||
| XM_017016205.1 | XP_016871694.1 | 7, 8 | 850 | |||
| GenBank mRNA | AF273055.1 | - | 7, 8 | 798 | ||
| BC052265.1 | - | 6, 7 | 453 | |||
| BC062300.1 | - | 6, 7 | 453 | |||
| BC096280.2 | - | 8, 9 | 599 | 40006844 | ||
| BC096281.2 | - | 7, 8 | 518 | 40006846 | ||
| X77567.1 | - | 7, 8 | 636 | |||
| Z31695.1 | - | 7, 8 | 491 | |||

INPP5A
Genomic Map