siRNA Details
| Gene Symbol: INPP5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321042.1 | NP_001307971.1 | 6 | 1037 | ||
| NM_005539.4 | NP_005530.3 | 9 | 984 | |||
| XM_017016204.1 | XP_016871693.1 | 8 | 638 | |||
| XM_017016205.1 | XP_016871694.1 | 9 | 979 | |||
| GenBank mRNA | AF273055.1 | - | 9 | 927 | ||
| BC052265.1 | - | 8 | 582 | |||
| BC062300.1 | - | 8 | 582 | |||
| BC096280.2 | - | 10 | 728 | 40006844 | ||
| BC096281.2 | - | 9 | 647 | 40006846 | ||
| X77567.1 | - | 9 | 765 | |||
| Z31695.1 | - | 9 | 620 | |||

INPP5A
Genomic Map