siRNA Details
| Gene Symbol: Stxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113569.1 | NP_001107041.1 | 16 | 1593 | ||
| NM_009295.2 | NP_033321.2 | 16 | 1588 | |||
| GenBank mRNA | AF326545.1 | - | 16 | 1447 | ||
| AF326563.1 | - | 16 | 1447 | |||
| AK004780.1 | - | 11 | 1312 | |||
| BC031728.1 | - | 16 | 1520 | 4480978 | ||
| D45903.1 | - | 16 | 1482 | |||

Stxbp1
Genomic Map