siRNA Details
| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011504.1 | NP_035634.1 | 18 | 1705 | ||
| GenBank mRNA | AK138609.1 | - | 18 | 1737 | ||
| AK138769.1 | - | 6 | 988 | |||
| AK151466.1 | - | 18 | 1700 | |||
| AK152261.1 | - | 18 | 1694 | |||
| AK155133.1 | - | 18 | 1703 | |||
| AK169799.1 | - | 16 | 1539 | |||
| BC062901.1 | - | 18 | 1714 | 6810254 | ||
| BC094055.1 | - | 18 | 1713 | |||
| D30798.1 | - | 18 | 1705 | |||
| U19521.1 | - | 18 | 1830 | |||

Stxbp3
Genomic Map