siRNA Details
| Gene Symbol: Supt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009297.2 | NP_033323.2 | 8, 9 | 1193 | ||
| GenBank mRNA | AK170533.1 | - | 8, 9 | 1192 | ||
| BC072657.1 | - | 8, 9 | 1113 | 30536338 | ||
| U40375.1 | - | 8, 9 | 1191 | |||

Supt6
Genomic Map