siRNA Details
| Gene Symbol: Surf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013678.2 | NP_038706.1 | 5 | 580 | ||
| GenBank mRNA | AK004931.1 | - | 1 | 2529 | ||
| AK157125.1 | - | 5 | 594 | |||
| AK165559.1 | - | 5 | 587 | |||
| AK172018.1 | - | 5 | 583 | |||
| AK177886.1 | - | 257 | ||||
| AK188020.1 | - | 3 | 257 | |||
| BC049779.1 | - | 4 | 1911 | |||
| BC096401.1 | - | 5 | 580 | 1365532 | ||

Surf2
Genomic Map