siRNA Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009302.3 | NP_033328.3 | 5 | 823 | ||
| GenBank mRNA | AF053974.1 | - | 5 | 715 | ||
| AK019882.1 | - | 5 | 767 | |||
| AK045295.1 | - | 5 | 795 | |||
| AK146773.1 | - | 5 | 815 | |||
| AK172990.1 | - | 5 | 760 | |||
| BC065136.1 | - | 5 | 765 | 30546814 | ||

Swap70
Genomic Map