siRNA Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009302.3 | NP_033328.3 | 4 | 644 | ||
| GenBank mRNA | AF053974.1 | - | 4 | 536 | ||
| AK019882.1 | - | 4 | 588 | |||
| AK041505.1 | - | 4 | 592 | |||
| AK146773.1 | - | 4 | 636 | |||
| BC065136.1 | - | 4 | 586 | 30546814 | ||

Swap70
Genomic Map