siRNA Details
| Gene Symbol: Sycp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011517.2 | NP_035647.2 | 7 | 676 | ||
| GenBank mRNA | AK131949.1 | - | 7 | 600 | ||
| BC048483.1 | - | 7 | 599 | |||
| BC089540.1 | - | 7 | 599 | |||
| BC099552.1 | - | 6 | 481 | |||
| BC132079.1 | - | 7 | 677 | 40130356 | ||
| BC138509.1 | - | 7 | 677 | 8861529 | ||
| Y08485.1 | - | 7 | 685 | |||

Sycp3
Genomic Map