siRNA Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110780.1 | NP_001104250.1 | 6, 7 | 974 | ||
| NM_013680.4 | NP_038708.3 | 6, 7 | 974 | |||
| GenBank mRNA | BC022954.1 | - | 6, 7 | 952 | 5368098 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syn1| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110780.1 | NP_001104250.1 | 6, 7 | 974 | ||
| NM_013680.4 | NP_038708.3 | 6, 7 | 974 | |||
| GenBank mRNA | BC022954.1 | - | 6, 7 | 952 | 5368098 | |