siRNA Details
| Gene Symbol: Sdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011520.3 | NP_035650.2 | 5 | 2143 | ||
| GenBank mRNA | AK087492.1 | - | 1 | 237 | ||
| AK129153.1 | - | 5 | 2143 | |||
| AK150120.1 | - | 3 | 1242 | |||
| AK155127.1 | - | 5 | 2128 | |||
| AK194562.1 | - | 1 | 166 | |||
| BC054795.1 | - | 5 | 2102 | 6834272 | ||
| BC059031.1 | - | 1 | 362 | |||
| BC062093.1 | - | 5 | 2039 | 5705893 | ||

Sdc3
Genomic Map