siRNA Details
| Gene Symbol: Syt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009307.3 | NP_033333.2 | 6 | 973 | ||
| GenBank mRNA | AF257304.1 | - | 5 | 728 | ||
| AK036357.1 | - | 6 | 974 | |||
| AK220537.1 | - | 3 | 287 | |||
| BC027019.1 | - | 5 | 734 | 5362352 | ||
| D37793.1 | - | 4 | 607 | |||

Syt2
Genomic Map