siRNA Details
| Gene Symbol: Syt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009307.3 | NP_033333.2 | 10 | 1472 | ||
| GenBank mRNA | AF257304.1 | - | 9 | 1227 | ||
| AK036357.1 | - | 10 | 1473 | |||
| AK220537.1 | - | 7 | 786 | |||
| BC027019.1 | - | 9 | 1233 | 5362352 | ||
| D37793.1 | - | 8 | 1106 | |||

Syt2
Genomic Map