siRNA Details
| Gene Symbol: T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161832.1 | NP_001155304.1 | 9 | 959 | ||
| GenBank mRNA | AF072546.1 | - | 6 | 551 | ||
| BC119261.1 | - | 3 | 317 | 8734010 | ||
| BC119287.1 | - | 3 | 317 | 8734036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

T2| Gene Symbol: T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161832.1 | NP_001155304.1 | 9 | 959 | ||
| GenBank mRNA | AF072546.1 | - | 6 | 551 | ||
| BC119261.1 | - | 3 | 317 | 8734010 | ||
| BC119287.1 | - | 3 | 317 | 8734036 | ||