siRNA Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009311.2 | NP_033337.1 | 2 | 149 | ||
| GenBank mRNA | BC117081.1 | - | 2 | 124 | 40126400 | |
| BC119122.1 | - | 2 | 124 | 8733871 | ||
| BC144737.1 | - | 2 | 124 | 9053285 | ||
| D17584.1 | - | 2 | 149 | |||

Tac1
Genomic Map