siRNA Details
| Gene Symbol: Tacr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009314.4 | NP_033340.3 | 2 | 831 | ||
| GenBank mRNA | AK033443.1 | - | 2 | 514 | ||
| BC106959.1 | - | 2 | 480 | 40039032 | ||
| X62933.1 | - | 2 | 831 | |||
More Information
Set Membership: |
|

Tacr2
Genomic Map