siRNA Details
| Gene Symbol: Tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253795.1 | NP_001240724.1 | 6 | 1016 | ||
| NM_009319.3 | NP_033345.2 | 8 | 1289 | |||
| GenBank mRNA | AK144205.1 | - | 6 | 854 | ||
| AK185093.1 | - | 1 | 113 | |||
| BC002028.1 | - | 8 | 1002 | 3489376 | ||
| U79962.1 | - | 8 | 1022 | |||

Tarbp2
Genomic Map