siRNA Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009320.4 | NP_033346.2 | 3, 4 | 535 | ||
| GenBank mRNA | AK041141.1 | - | 3, 4 | 508 | ||
| AK143510.1 | - | 3, 4 | 499 | |||
| AK162776.1 | - | 3, 4 | 499 | |||
| AK214888.1 | - | 1, 2 | 50 | |||
| BB636410.1 | - | 3, 4 | 508 | |||
| BC015245.1 | - | 3, 4 | 472 | 4242193 | ||
| L03292.2 | - | 3, 4 | 507 | |||

Slc6a6
Genomic Map