siRNA Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020601.2 | NP_065626.1 | 9 | 1020 | ||
| GenBank mRNA | AK030547.1 | - | 9 | 1235 | ||
| AK031937.1 | - | 5 | 481 | |||
| AK036996.1 | - | 8 | 1464 | |||
| AK038674.1 | - | 9 | 1021 | |||
| BC043105.1 | - | 9 | 1143 | 6406579 | ||

Tbl1x
Genomic Map