siRNA Details
| Gene Symbol: Tbrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130457.1 | NP_001123929.1 | 10, 11 | 1905 | ||
| NM_134011.2 | NP_598772.1 | 10, 11 | 1905 | |||
| GenBank mRNA | AK028966.1 | - | 10, 11 | 1836 | ||
| AK075897.1 | - | 7, 8 | 1179 | |||
| AK129245.1 | - | 10, 11 | 1860 | |||
| AK144813.1 | - | 10, 11 | 1904 | |||
| AK166760.1 | - | 11, 12 | 1813 | |||
| AK168088.1 | - | 10, 11 | 1903 | |||
| AK181021.1 | - | 3, 4 | 419 | |||
| BC016483.1 | - | 10, 11 | 1861 | 3155271 | ||
| BC031155.1 | - | 10, 11 | 1876 | 4976690 | ||
| BC031910.1 | - | 10, 11 | 1875 | 5362943 | ||

Tbrg4
Genomic Map