siRNA Details
| Gene Symbol: Tbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009324.2 | NP_033350.2 | 2 | 1139 | ||
| GenBank mRNA | AK080703.1 | - | 1 | 781 | ||
| AK137225.1 | - | 2 | 345 | |||
| AK149348.1 | - | 2 | 947 | |||
| U15566.1 | - | 2 | 675 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx2| Gene Symbol: Tbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009324.2 | NP_033350.2 | 2 | 1139 | ||
| GenBank mRNA | AK080703.1 | - | 1 | 781 | ||
| AK137225.1 | - | 2 | 345 | |||
| AK149348.1 | - | 2 | 947 | |||
| U15566.1 | - | 2 | 675 | |||