siRNA Details
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011540.2 | NP_035670.2 | 2 | 792 | ||
| GenBank mRNA | AJ223854.1 | - | 2 | 792 | ||
| AK010167.1 | - | 2 | 780 | |||
| Y15845.1 | - | 2 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcap| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011540.2 | NP_035670.2 | 2 | 792 | ||
| GenBank mRNA | AJ223854.1 | - | 2 | 792 | ||
| AK010167.1 | - | 2 | 780 | |||
| Y15845.1 | - | 2 | 782 | |||