siRNA Details
| Gene Symbol: Hnf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009327.3 | NP_033353.2 | 2 | 573 | ||
| GenBank mRNA | AK141225.1 | - | 2 | 380 | ||
| AK144051.1 | - | 2 | 580 | |||
| BC080698.1 | - | 2 | 575 | 30471380 | ||
| M57966.1 | - | 553 | ||||

Hnf1a
Genomic Map