siRNA Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009329.3 | NP_033355.2 | 5 | 791 | ||
| GenBank mRNA | AB221586.1 | - | 4 | 595 | ||
| AF184111.1 | - | 5 | 753 | |||
| AK030469.1 | - | 5 | 770 | |||
| BC050843.1 | - | 5 | 791 | 6506256 | ||
| BC087540.1 | - | 5 | 927 | 4239789 | ||
| L77247.1 | - | 4 | 655 | |||

Zfp354a
Genomic Map