siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083967.1 | NP_001077436.1 | 15 | 1431 | ||
| NM_013685.2 | NP_038713.1 | 15 | 1746 | |||
| GenBank mRNA | AK014343.1 | - | 8 | 815 | ||
| AK028748.1 | - | 15 | 1357 | |||
| AK081012.1 | - | 15 | 1383 | |||
| AK132140.1 | - | 8 | 772 | |||
| AK133885.1 | - | 16 | 2158 | |||
| AK151433.1 | - | 8 | 717 | |||
| BC014293.1 | - | 8 | 768 | 4014231 | ||
| BC043050.1 | - | 14 | 1206 | 5708370 | ||
| S75870.1 | - | 15 | 1318 | |||
| U16321.1 | - | 9 | 805 | |||
| U16322.1 | - | 16 | 1718 | |||
| X91753.1 | - | 15 | 1370 | |||

Tcf4
Genomic Map