siRNA Details
| Gene Symbol: Zeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011546.3 | NP_035676.1 | 6 | 1252 | ||
| GenBank mRNA | AK140038.1 | - | 6 | 1415 | ||
| AK141437.1 | - | 2 | 582 | |||
| BC090987.1 | - | 6 | 1218 | |||
| BC139768.1 | - | 6 | 1221 | 40111224 | ||
| BC139769.1 | - | 6 | 1221 | 40111227 | ||
| D76432.1 | - | 6 | 1221 | |||
| U26259.1 | - | 6 | 1238 | |||

Zeb1
Genomic Map