siRNA Details
| Gene Symbol: PDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000209.3 | NP_000200.1 | 2 | 1454 | ||
| GenBank mRNA | U30329.1 | - | 2 | 1366 | ||
| X99894.1 | - | 2 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDX1| Gene Symbol: PDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000209.3 | NP_000200.1 | 2 | 1454 | ||
| GenBank mRNA | U30329.1 | - | 2 | 1366 | ||
| X99894.1 | - | 2 | 1445 | |||