siRNA Details
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011549.3 | NP_035679.3 | 1 | 166 | ||
| GenBank mRNA | AF079095.1 | - | 1 | 147 | ||
| AK146016.1 | - | 1 | 191 | |||
| BC064789.1 | - | 1 | 166 | 30429545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfeb| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011549.3 | NP_035679.3 | 1 | 166 | ||
| GenBank mRNA | AF079095.1 | - | 1 | 147 | ||
| AK146016.1 | - | 1 | 191 | |||
| BC064789.1 | - | 1 | 166 | 30429545 | ||