siRNA Details
| Gene Symbol: Tcte2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022311.2 | NP_071706.2 | 3 | 240 | ||
| GenBank mRNA | AK008572.1 | - | 226 | |||
| BC049646.1 | - | 4 | 294 | |||
| BC092528.1 | - | 3 | 240 | 6741782 | ||
| U46151.1 | - | 4 | 707 | |||

Tcte2
Genomic Map