siRNA Details
| Gene Symbol: ITGA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000210.3 | NP_000201.2 | 22 | 3099 | ||
| NM_001079818.2 | NP_001073286.1 | 22 | 3099 | |||
| NM_001316306.1 | NP_001303235.1 | 22 | 2785 | |||
| XM_006712510.1 | XP_006712573.1 | 22 | 3023 | |||
| XM_006712511.1 | XP_006712574.1 | 22 | 3023 | |||
| XM_017004005.1 | XP_016859494.1 | 22 | 2795 | |||
| XM_017004006.1 | XP_016859495.1 | 22 | 2750 | |||
| XM_017004007.1 | XP_016859496.1 | 22 | 2795 | |||
| XM_017004008.1 | XP_016859497.1 | 22 | 2750 | |||
| GenBank mRNA | AB208842.1 | - | 22 | 2766 | ||
| AK294436.1 | - | 18 | 2187 | |||
| AK296496.1 | - | 22 | 2721 | |||
| BC050585.1 | - | 16 | 1705 | |||
| BC136455.1 | - | 22 | 2973 | 9020444 | ||
| BC136456.1 | - | 22 | 2973 | 9020445 | ||
| S66196.1 | - | 1 | 88 | |||
| S66213.1 | - | 1 | 88 | |||
| X53586.1 | - | 22 | 3011 | |||
| X59512.1 | - | 22 | 2847 | |||

ITGA6
Genomic Map