siRNA Details
| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009347.2 | NP_033373.2 | 20 | 6238 | ||
| GenBank mRNA | AK136719.1 | - | 5 | 1064 | ||
| BC138426.1 | - | 20 | 6021 | 8861446 | ||
| BC171953.1 | - | 20 | 6021 | 9054239 | ||
| X99805.1 | - | 20 | 6238 | |||

Tecta
Genomic Map