siRNA Details
| Gene Symbol: Tef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017376.3 | NP_059072.1 | 4 | 868 | ||
| NM_153484.3 | NP_705617.2 | 4 | 775 | |||
| GenBank mRNA | AF194420.1 | - | 4 | 802 | ||
| AK042146.1 | - | 4 | 1029 | |||
| AK141606.1 | - | 4 | 696 | |||
| AK145125.1 | - | 4 | 837 | |||
| AK147965.1 | - | 4 | 801 | |||
| AK154472.1 | - | 4 | 813 | |||
| AY540631.1 | - | 4 | 854 | |||
| AY540632.1 | - | 4 | 669 | |||
| BC017689.1 | - | 4 | 631 | 4242534 | ||
| BC036982.1 | - | 4 | 832 | 4976241 | ||

Tef
Genomic Map