siRNA Details
| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009349.3 | NP_033375.1 | 3 | 527 | ||
| GenBank mRNA | AK002281.1 | - | 3 | 575 | ||
| AK013010.1 | - | 3 | 564 | |||
| BC013518.1 | - | 3 | 553 | 4236077 | ||
| M88694.1 | - | 3 | 528 | |||

Inmt
Genomic Map