siRNA Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286628.1 | NP_001273557.1 | 1, 2 | 319 | ||
| NM_009352.3 | NP_033378.1 | 1, 2 | 319 | |||
| GenBank mRNA | AB109563.1 | - | 1, 2 | 299 | ||
| AK021235.1 | - | 1, 2 | 309 | |||
| AK132022.1 | - | 1, 2 | 302 | |||
| BB652245.1 | - | 1, 2 | 320 | |||
| BC034866.1 | - | 1, 2 | 307 | 3470277 | ||
| U65586.1 | - | 1, 2 | 299 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001012464, XM_006237729, BC089888.

Terf1
Genomic Map