siRNA Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011571.3 | NP_035701.3 | 3, 4 | 1531 | ||
| GenBank mRNA | AB003494.1 | - | 3, 4 | 1869 | ||
| AK144302.1 | - | 3, 4 | 1114 | |||
| BC099699.1 | - | 3, 4 | 1531 | 6821934 | ||

Tesk1
Genomic Map