siRNA Details
| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198292.3 | NP_938034.2 | 4 | 2008 | ||
| GenBank mRNA | AK029537.1 | - | 4 | 2016 | ||
| AK129434.1 | - | 4 | 2008 | |||
| AK168397.1 | - | 4 | 2018 | |||
| BC057406.1 | - | 3 | 418 | 5687258 | ||

Tex2
Genomic Map