siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009357.2 | NP_033383.1 | 2 | 181 | ||
| GenBank mRNA | AK149789.1 | - | 2 | 146 | ||
| AK157834.1 | - | 2 | 160 | |||
| AK182876.1 | - | 1 | 21 | |||
| AK201329.1 | - | 2 | 78 | |||
| BC003802.1 | - | 2 | 125 | 3486968 | ||
| BY250528.1 | - | 2 | 182 | |||
| X80426.1 | - | 2 | 129 | |||
| X81058.1 | - | 2 | 127 | |||

Tex261
Genomic Map