siRNA Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081654.2 | NP_001075123.1 | 6 | 1011 | ||
| NM_001286498.1 | NP_001273427.1 | 6 | 1211 | |||
| NM_011573.3 | NP_035703.2 | 6 | 1466 | |||
| GenBank mRNA | AK003113.1 | - | 7 | 972 | ||
| AK009326.1 | - | 6 | 1317 | |||
| AK159046.1 | - | 6 | 1717 | |||
| BC002248.1 | - | 6 | 975 | 3493141 | ||
| BC006608.1 | - | 6 | 949 | 3157453 | ||
| DV656490.1 | - | 5 | 664 | |||

Tex264
Genomic Map