siRNA Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081654.2 | NP_001075123.1 | 5 | 751 | ||
| NM_001286498.1 | NP_001273427.1 | 5 | 951 | |||
| NM_011573.3 | NP_035703.2 | 5 | 1206 | |||
| GenBank mRNA | AK003113.1 | - | 6 | 712 | ||
| AK009326.1 | - | 5 | 1057 | |||
| DV656490.1 | - | 4 | 404 | |||
| DV661702.1 | - | 3 | 526 | |||

Tex264
Genomic Map